missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloroform-d, 99.8% (Isotopic)

NMR solvent

Manufacturer:  Alfa Aesar™ 8954130

 View more versions of this product

Catalog No. AA8954130



Specifications

Specifications

Chloroform-d
1.4445
62°C
-64°C
Isotopic
CHCl3
UN1888
HEDRZPFGACZZDS-MICDWDOJSA-N
trichloro(deuterio)methane
71583
120.38
99.8%
1.50g/mL
Moisture & Light Sensitive
Colorless
250g
865-49-6
MFCD00000827
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
C(Cl)(Cl)Cl
120.375
CHEBI:85365
Liquid

Chemical Identifiers

865-49-6
120.375
HEDRZPFGACZZDS-MICDWDOJSA-N
71583
trichloro(deuterio)methane
CHCl3
MFCD00000827
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
CHEBI:85365
C(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H351-H373-H302-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
P260-P280-P281-P362-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROFORM

Recommended Storage : 2°C to 8°C

SDS Documents