Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloroform-d, 100% (Isotopic)

Chemical Identifiers

Chemical Name or Material Chloroform-d
CAS 865-49-6
Name Note Isotopic
Formula Weight 120.38
MDL Number MFCD00000827
UN Number UN1888
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42270KV

  SDS
alfa aesar™
42270KV
10 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid In Prescored Ampoules, (0.75Ml/ampoule)
Formula Weight 120.38
Sensitivity Moisture & Light Sensitive
Melting Point −64°C
Color Colorless
Refractive Index 1.4445
Health Hazard 3 GHS P Statement
P260-P280-P281-P362-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2°C to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROFORM
Chemical Name or Material Chloroform-d
Density 1.50g/mL
Name Note Isotopic
Boiling Point 62°C
Assay 100%
Quantity 10 each
MDL Number MFCD00000827
CAS 865-49-6
UN Number UN1888
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H373-H302-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Safety and Handling

Warning

GHS P Statement
P260-P280-P281-P362-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H373-H302-H315
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.

2°C to 8°C