Please login to your online account to display your discounted pricing

Chloroform ChromAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Chloroform
CAS 67-66-3
Molecular Formula CHCl3
Formula Weight 119.38
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK44431

 
avantor performance
444306
Glass 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK44434

 
avantor performance
444310
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MKH4074

 
avantor performance
H40710
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Chloroform
CAS 67-66-3
Molecular Formula CHCl3
Formula Weight 119.38
Packaging Glass
Quantity 1L