missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform - Butanol Reagent, Ricca Chemical

Chemical Identifiers

CAS 67-66-3,71-36-3,7732-18-5,7732-18-5 (Water, Deionized)
CAS Min % 74.0,24.4,0.05
CAS Max % 77.0,25.4,0.05
Physical Form Liquid
Chemical Name or Material Chloroform - Butanol Reagent
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
19961 Ricca Chemical Company
RI19961
Glass amber 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
199632 Ricca Chemical Company
RI199632
Glass amber 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

67-66-3,71-36-3,7732-18-5,7732-18-5 (Water, Deionized)
77.0,25.4,0.05
Chloroform - Butanol Reagent
74.0,24.4,0.05
Liquid

Specifications

67-66-3,71-36-3,7732-18-5,7732-18-5 (Water, Deionized)
77.0,25.4,0.05
Glass amber
4L
74.0,24.4,0.05
Liquid
Clear colorless
Chloroform - Butanol Reagent