Please login to your online account to display your discounted pricing

Chloroform-Butanol Reagent, For Organic Acids, LabChem™

$199.91 - $199.91

Chemical Identifiers

Chemical Name or Material Chloroform-Butanol Reagent
CAS 67-66-3
CAS Max % 83.754
CAS 71-36-3
CAS Max % 15.4
CAS 64-17-5
CAS Max % 0.846
Name Note For Organic Acids
Linear Formula CHCl3
Formula Weight 119.38
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC130502

 
lab chem
LC130502
Amber Glass 1L Each for $199.91
Description & Specifications

Specifications

Formula Weight 119.38
Chemical Name or Material Chloroform-Butanol Reagent
Identification Passes Test
CAS Max % 0.846
Packaging Amber Glass
Specific Gravity 1.3g/mL
Storage Information Room Temperature
Name Note For Organic Acids
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Physical Form Liquid
Color Clear, colorless
Quantity 1L
Linear Formula CHCl3
CAS 67-66-3
UN Number UN2810
Health Hazard 1 Danger
CAS Max % 83.754
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
Causes damage to organs (liver, kidneys, central nervous system, hearing organ) through prolonged or repeated exposure (inhalation, oral).
Toxic to aquatic life.
Density 1.3g/mL
CAS 71-36-3
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 15.4
CAS 64-17-5
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
Causes damage to organs (liver, kidneys, central nervous system, hearing organ) through prolonged or repeated exposure (inhalation, oral).
Toxic to aquatic life.