Please login to your online account to display your discounted pricing

Chloroform, Anhydrous DriSolv™, EMD Millipore

Septum-Sealed Bottle

$151.54 - $151.54

Chemical Identifiers

Chemical Name or Material Chloroform, Anhydrous
CAS 67-66-3
Molecular Formula CHCl3
Formula Weight 119.38
UN Number 1888
Synonym Trichloromethane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MCX10566

 
emd millipore corporation
CX10566
Glass Bottle 1L Each for $151.54
Description & Specifications

Specifications

Boiling Point 60.5°C
Formula Weight 119.38
Melting Point −63°C
Assay ≥99.8%
Quantity 1L
Storage Information Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
CAS 67-66-3
Chemical Name or Material Chloroform, Anhydrous
UN Number 1888
Physical Form Liquid
Synonym Trichloromethane
Odor Pleasant. Ethereal
Packaging Glass Bottle
Color Colorless
Vapor Pressure 22.3 kPa (167mm Hg) [20°C]
Molecular Formula CHCl3