Please login to your online account to display your discounted pricing

Chloroform ACS AR, Macron Fine Chemicals™


macron

Quantity 650lb.
Packaging Lined Steel Dr

Manufacturer: avantor performance  444024

 View more versions of this product

Catalog No. MK4440650

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 119.38
Purity Grade ACS Reagent Grade
Quantity 650lb.
Packaging Lined Steel Dr
Molecular Formula CHCl3
CAS 67-66-3
Chemical Name or Material Chloroform

Specifications

Chemical Name or Material Chloroform
CAS 67-66-3
Purity Grade ACS Reagent Grade
Molecular Formula CHCl3
Formula Weight 119.38