Please login to your online account to display your discounted pricing

Chloroform, 99+%, extra pure, stabilized with amylene, ACROS Organics™


Quantity 5L

Manufacturer: thermo fisher scientific  AC383760050

 View more versions of this product

Catalog No. AC383760050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 119.38
Purity Grade Extra Pure
Assay ≥99.0%
Quantity 5L
Color Clear
Merck Index 15, 2142
Density 1.4800g/mL
Linear Formula CHCl3
Water 0.1% max. (K.F.)
CAS 513-35-9
Stabilizer approx. 100ppm amylene
Residue after Evaporation 0.002% max.
Name Note Stabilized with amylene
Health Hazard 1 Warning
Chemical Name or Material Chloroform
Molecular Formula CHCl3
Assay Percent Range 99+%
CAS 67-66-3
MDL Number MFCD00000826
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
Health Hazard 3 GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Specifications

Chemical Name or Material Chloroform
Name Note Stabilized with amylene
CAS 67-66-3
CAS 513-35-9
Purity Grade Extra Pure
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula CHCl3
Linear Formula CHCl3
Formula Weight 119.38
MDL Number MFCD00000826
Accessories

Warning

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

SDS & Safety Documents