Please login to your online account to display your discounted pricing

Chloroform, +99%, for analysis, stabilized with ethanol, ACROS Organics™


Quantity 10L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC232090100

 View more versions of this product

Catalog No. AC232090100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 119.38
Assay ≥99.0%
Quantity 10L
Color Clear
Packaging Plastic Jerrycan
Purity Grade For analysis
Density 1.4800g/mL
Beilstein 01, 61
Merck Index 15, 2142
Fieser 01,130; 08,92; 15,84
Nickel (Ni) 0.1ppm max.
Iron (Fe) 0.5ppm max.
CAS 64-17-5
Trace Analysis 0.5ppm max.
Stabilizer approx. 1% ethanol
Cobalt (Co) 0.1ppm max.
MDL Number MFCD00000826
Chemical Name or Material Chloroform
Assay Percent Range 99+%
Molecular Formula CHCl3
Cadmium (Cd) 0.1ppm max.
Free Chlorine Passes Test
Aldehyde and Ketone 0.005% max. (as acetone)
Water 0.01% max.
Residue after Evaporation 0.001% max.
Copper (Cu) 0.1ppm max.
Hydrochloric Acid 0.001% max.
Linear Formula CHCl3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
CAS 67-66-3
Health Hazard 2 GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

Specifications

Chemical Name or Material Chloroform
CAS 67-66-3
CAS 64-17-5
Purity Grade For analysis
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula CHCl3
Linear Formula CHCl3
Formula Weight 119.38
MDL Number MFCD00000826

For analysis

Accessories

Warning

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

SDS & Safety Documents