Please login to your online account to display your discounted pricing

Chloroform, +99%, for analysis, stabilized with ethanol, ACROS Organics™

$108.42 - $205.93

Chemical Identifiers

Chemical Name or Material Chloroform
CAS 67-66-3
CAS 64-17-5
Purity Grade For analysis
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula CHCl3
Linear Formula CHCl3
Formula Weight 119.38
MDL Number MFCD00000826
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC232090100

  SDS
thermo fisher scientific
AC232090100
Plastic Jerrycan 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC232090025

  SDS
thermo fisher scientific
232090025
Glass Bottle 2.5L Each for $205.93

AC232090010

  SDS
thermo fisher scientific
232090010
Glass Bottle 1L Each for $108.42

AC232090250

  SDS
thermo fisher scientific
AC232090250
Metal drum 25L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥99.0%
Formula Weight 119.38
Quantity 10L
Color Clear
Packaging Plastic Jerrycan
Purity Grade For analysis
Density 1.4800g/mL
Beilstein 01, 61
Fieser 01,130; 08,92; 15,84
Merck Index 15, 2142
Iron (Fe) 0.5ppm max.
Nickel (Ni) 0.1ppm max.
Chemical Name or Material Chloroform
Residue after Evaporation 0.001% max.
Stabilizer approx. 1% ethanol
Aldehyde and Ketone 0.005% max. (as acetone)
Trace Analysis 0.5ppm max.
Water 0.01% max.
Assay Percent Range 99+%
Molecular Formula CHCl3
Linear Formula CHCl3
MDL Number MFCD00000826
Cadmium (Cd) 0.1ppm max.
Cobalt (Co) 0.1ppm max.
Copper (Cu) 0.1ppm max.
CAS 64-17-5
Free Chlorine Passes Test
Hydrochloric Acid 0.001% max.
CAS 67-66-3
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
Health Hazard 3 GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

For analysis

Safety and Handling

Warning

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.