Please login to your online account to display your discounted pricing

Chloroform 99%, ACROS Organics™


Quantity 10L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC158210100

 View more versions of this product

Catalog No. AC158210100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0% (excluding stabilizer)
Formula Weight 119.38
Purity Grade Extra Pure
Physical Form Liquid
Quantity 10L
Color Clear
Beilstein 01, 61
Merck Index 15, 2142
Packaging Plastic Jerrycan
Density 1.4800g/mL
Decomposition Information 290°C
Fieser 01,130; 08,92; 15,84
Linear Formula CHCl3
CAS 64-17-5
Water 0.02% max. (K.F.)
CAS 67-66-3
Chemical Name or Material Chloroform
MDL Number MFCD00000826
Residue after Evaporation 0.002% max.
Additional Information Vapor Pressure: 213mbar at 20°C
Stabilizer Approx. 1% ethanol
Health Hazard 1 Warning
Assay Percent Range 99+%
Health Hazard 3 GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Molecular Formula CHCl3
Health Hazard 2 GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

Specifications

Chemical Name or Material Chloroform
CAS 67-66-3
CAS 64-17-5
Purity Grade Extra Pure
Assay ≥99.0% (excluding stabilizer)
Assay Percent Range 99+%
Molecular Formula CHCl3
Linear Formula CHCl3
Formula Weight 119.38
MDL Number MFCD00000826
Accessories

Warning

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

SDS & Safety Documents