missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, 99+%, Acros Organics™

Manufacturer:  Acros Organics 158210025

 View more versions of this product

Catalog No. AC158210025


 

Specifications

Specifications

Chloroform
Vapor Pressure: 213mbar at 20°C
≥99%
CHCl3
MFCD00000826
01,130; 08,92; 15,84
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
C(Cl)(Cl)Cl
119.369
CHEBI:35255
Liquid
Glass Bottle
0.002% max.
1.48
0.02% max. (K.F.)
290°C
Undesignated
2.5L
67-66-3,64-17-5
Extra Pure
99+%
CHCl3
01, 61
15, 2142
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
1.4800g/mL
1.4443 to 1.4463
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
Approx. 1% ethanol
0.56 mPa.s (20°C)
61.0°C
-63.0°C

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
CHEBI:35255
CHCl3
MFCD00000826
6212
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

SDS Documents