missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chloroform, 99.8% min., ACS, MilliporeSigma™
Supplier: Technidata CX10552
Specifications
Chloroform | |
67-66-3 | |
1 L | |
99.8% min. | |
MFCD00000826 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
6212 | |
119.38 |
For ACS analysis | |
Colorless | |
Liquid | |
CHCl3 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 |
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
trichloromethane |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 | |
ClC(Cl)Cl |