Learn More
chlorodiphenylphosphine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 347300050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chlorodiphenylphosphine | |
Colorless to Yellow | |
320.0°C | |
Authentic | |
Glass bottle | |
1.6340 to 1.6370 | |
MFCD00000529 | |
16, 763 | |
1.229 | |
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane | |
XGRJZXREYAXTGV-UHFFFAOYSA-N | |
chloro(diphenyl)phosphane | |
66180 | |
98% |
1079-66-9 | |
1.2290g/mL | |
138°C | |
97.5% min. (GC) | |
C12H10ClP | |
(C6H5)2PCl | |
5 g | |
11,120; 16,77 | |
15, 2139 | |
Solubility in water: reacts violently with water | |
ClP(C1=CC=CC=C1)C1=CC=CC=C1 | |
220.64 | |
220.64 | |
Liquid |
Chemical Identifiers
1079-66-9 | |
220.64 | |
XGRJZXREYAXTGV-UHFFFAOYSA-N | |
66180 | |
ClP(C1=CC=CC=C1)C1=CC=CC=C1 |
C12H10ClP | |
MFCD00000529 | |
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane | |
chloro(diphenyl)phosphane |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 214-093-2
Recommended Storage : may become turbid on storage
RUO – Research Use Only