Please login to your online account to display your discounted pricing

Chlorodiphenylphosphine, 95%, tech., ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC109135000

 View more versions of this product

Catalog No. AC109135000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Technical Grade
Formula Weight 220.64
Quantity 500mL
Density 1.2290g/mL
Beilstein 16, 763
Merck Index 15, 2139
Fieser 11,120; 16,77
Packaging Glass bottle
Linear Formula (C6H5)2PCl
MDL Number MFCD00000529
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 1079-66-9
Assay Percent Range 95%
Molecular Formula C12H10ClP
Chemical Name or Material Chlorodiphenylphosphine

Specifications

Chemical Name or Material Chlorodiphenylphosphine
CAS 1079-66-9
Purity Grade Technical Grade
Assay Percent Range 95%
Molecular Formula C12H10ClP
Linear Formula (C6H5)2PCl
Formula Weight 220.64
MDL Number MFCD00000529
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with water liberates toxic gas.

SDS & Safety Documents