Learn More
Chlorodiphenylphosphine, 95%, tech., Thermo Scientific Chemicals
$77.77 - $77.77
Chemical Identifiers
CAS | 1079-66-9 |
---|---|
Molecular Formula | C12H10ClP |
Molecular Weight (g/mol) | 220.64 |
MDL Number | MFCD00000529 |
InChI Key | XGRJZXREYAXTGV-UHFFFAOYSA-N |
Synonym | chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane |
PubChem CID | 66180 |
SMILES | ClP(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC109130250
|
Thermo Scientific Chemicals
109130250 |
25 mL | Glass bottle |
Each for $77.77
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
1079-66-9 | |
220.64 | |
XGRJZXREYAXTGV-UHFFFAOYSA-N | |
66180 |
C12H10ClP | |
MFCD00000529 | |
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane | |
ClP(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
1079-66-9 | |
320.0°C | |
94% min. (GC) | |
C12H10ClP | |
(C6H5)2PCl | |
25 mL | |
11,120; 16,77 | |
15, 2139 | |
Solubility in water: reacts violently with water | |
ClP(C1=CC=CC=C1)C1=CC=CC=C1 | |
220.64 | |
220.64 | |
Technical |
1.2290g/mL | |
138°C | |
Glass bottle | |
1.6340 to 1.6370 | |
MFCD00000529 | |
16, 763 | |
1.229 | |
chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane | |
XGRJZXREYAXTGV-UHFFFAOYSA-N | |
chlorodiphenylphosphane | |
66180 | |
95% | |
Chlorodiphenylphosphine |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 214-093-2
RUO – Research Use Only