Please login to your online account to display your discounted pricing

chlorodimethylthexylsilane, 95%, ACROS Organics™

$32.65 - $81.76

Chemical Identifiers

Chemical Name or Material chlorodimethylthexylsilane
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H19ClSi
Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl
Formula Weight 178.78
MDL Number MFCD00012175
Synonym Dimethylthexylsilyl chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC408601000

  SDS
thermo fisher scientific
AC408601000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC408600250

  SDS
thermo fisher scientific
408600250
Glass bottle 25g Each for $81.76

AC408600050

  SDS
thermo fisher scientific
408600050
Glass bottle 5g Each for $32.65
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 178.78
Quantity 100g
Color Clear colorless to almost colorless
Density 0.9g/mL
Chemical Name or Material chlorodimethylthexylsilane
Molecular Formula C8H19ClSi
Synonym Dimethylthexylsilyl chloride
Refractive Index 1.4480 to 1.4500 (20°C, 589nm)
Assay Percent Range 95%
MDL Number MFCD00012175
CAS 67373-56-2
Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.