Please login to your online account to display your discounted pricing

chlorodimethylthexylsilane, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  408600250

 View more versions of this product

Catalog No. AC408600250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Liquid
Formula Weight 178.78
Quantity 25g
Color Clear colorless to almost colorless
Density 0.9g/mL
Refractive Index 1.4480 to 1.4500 (20°C, 589nm)
Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl
Synonym Dimethylthexylsilyl chloride
CAS 67373-56-2
Molecular Formula C8H19ClSi
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00012175
Packaging Glass bottle
Infrared Spectrum Authentic
Chemical Name or Material chlorodimethylthexylsilane
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

Specifications

Chemical Name or Material chlorodimethylthexylsilane
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H19ClSi
Linear Formula (CH3)2CHC(CH3)2Si(CH3)2Cl
Formula Weight 178.78
MDL Number MFCD00012175
Synonym Dimethylthexylsilyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents