missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

chlorodimethylsilane, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC162842500

 View more versions of this product

Catalog No. AC162842500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Reduction reagent
Specifications

Specifications

chlorodimethylsilane
-28°C
Glass bottle
0.85
Colorless
250g
96%
(CH3)2SiHCl
13,73
Solubility in water: reacts
C[Si](C)Cl
94.62
94.62
96%
0.8500g/mL
Authentic
1.3820 to 1.384
34.6°C
-111°C
1066-35-9
C2H7ClSi
MFCD00000495
chloro dimethyl silicon
QABCGOSYZHCPGN-UHFFFAOYSA-N
chloro(dimethyl)silicon
6327132
Liquid

Chemical Identifiers

1066-35-9
94.62
QABCGOSYZHCPGN-UHFFFAOYSA-N
6327132
C[Si](C)Cl
C2H7ClSi
MFCD00000495
chloro dimethyl silicon
chloro(dimethyl)silicon
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Extremely flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 213-912-

SDS Documents