missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorodimethylphenylsilane, 97+%

Manufacturer:  Alfa Aesar™ A1563822

 View more versions of this product

Catalog No. AAA1563822



Description

Description

Applications
Chlorodimethylphenylsilane is used in the synthesis of enantioenriched allenylsilanes through an ortho-ester Claisen rearrangement of chiral and silylpropargylic alcohols.

Solubility
Miscible with tetrahydrofuran, ether and hexane.

Notes
Incompatible with strong oxidizing agents, acids, bases, and moisture.
Specifications

Specifications

Chlorodimethylphenylsilane
C8H11ClSi
UN2987
chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
C[Si](C)(C1=CC=CC=C1)Cl
170.711
170.72
192°C to 193°C
72°C (161°F)
768-33-2
MFCD00000499
606292
KWYZNESIGBQHJK-UHFFFAOYSA-N
chloro-dimethyl-phenylsilane
13029
≥97%
100g

Chemical Identifiers

768-33-2
170.711
KWYZNESIGBQHJK-UHFFFAOYSA-N
13029
C[Si](C)(C1=CC=CC=C1)Cl
C8H11ClSi
MFCD00000499
chlorodimethylphenylsilane, chlorodimethyl phenyl silane, phenyldimethylchlorosilane, silane, chlorodimethylphenyl, dimethylphenylchlorosilane, chloro dimethyl phenylsilane, phenyl dimethylchlorosilane, chloro-dimethyl-phenyl-silane, benzene, chlorodimethylsilyl, pubchem10814
chloro-dimethyl-phenylsilane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

TSCA : TSCA

Documents