Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorodimethyl-n-propylsilane, 97%

Chemical Identifiers

Chemical Name or Material Chlorodimethyl-n-propylsilane
CAS 17477-29-1
Molecular Formula C5H13ClSi
Formula Weight 136.7
MDL Number MFCD00039301
UN Number UN2985
Beilstein 1732132
Synonym Dimethyl-n-propylsilyl chloride; n-Propyldimethylchlorosilane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1662106

  SDS
alfa aesar™
L1662106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1662114

  SDS
alfa aesar™
L1662114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 136.7
Assay 97%
Molecular Formula C5H13ClSi
Chemical Name or Material Chlorodimethyl-n-propylsilane
MDL Number MFCD00039301
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
CAS 17477-29-1
Flash Point 8°C (46°F)
TSCA TSCA
UN Number UN2985
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 5g
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Beilstein 1732132
Boiling Point 113° to 114°C
Synonym Dimethyl-n-propylsilyl chloride; n-Propyldimethylchlorosilane
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.