Learn More
Chlorodimethyl-n-octylsilane, 97%, Thermo Scientific Chemicals
$47.67 - $47.67
Chemical Identifiers
CAS | 18162-84-0 |
---|---|
Molecular Formula | C10H23ClSi |
Molecular Weight (g/mol) | 206.829 |
MDL Number | MFCD00000511 |
InChI Key | DBKNGKYVNBJWHL-UHFFFAOYSA-N |
Synonym | n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc |
PubChem CID | 87482 |
IUPAC Name | chloro-dimethyl-octylsilane |
SMILES | CCCCCCCC[Si](C)(C)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1656406
|
Thermo Scientific Chemicals
L1656406 |
5 g |
Each for $47.67
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18162-84-0 | |
206.829 | |
DBKNGKYVNBJWHL-UHFFFAOYSA-N | |
87482 | |
CCCCCCCC[Si](C)(C)Cl |
C10H23ClSi | |
MFCD00000511 | |
n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc | |
chloro-dimethyl-octylsilane |
Specifications
222°C to 225°C | |
5 g | |
MFCD00000511 | |
1738376 | |
n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc | |
CCCCCCCC[Si](C)(C)Cl | |
206.829 | |
206.83 | |
Acrid | |
Moisture sensitive | |
Chlorodimethyl-n-octylsilane |
18162-84-0 | |
C10H23ClSi | |
UN2987 | |
92°C (198°F) | |
DBKNGKYVNBJWHL-UHFFFAOYSA-N | |
chloro-dimethyl-octylsilane | |
87482 | |
97% | |
1.435 | |
0.873 |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber : 242-044-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only