Learn More
chlorodiisopropylphosphine, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 321550250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chlorodiisopropylphosphine | |
Yellow | |
69.0°C (33.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4740 to 1.4760 | |
MFCD00015027 | |
0.95 | |
JZPDBTOWHLZQFC-UHFFFAOYSA-N | |
chloro-di(propan-2-yl)phosphane | |
538967 | |
96% |
40244-90-4 | |
0.9500g/mL | |
3°C | |
96% | |
C6H14ClP | |
[(CH3)2CH]2PCl | |
25 g | |
chlorodiisopropylphosphine, diisopropylchlorophosphine, di-i-propylchlorophosphine, phosphinous chloride, bis 1-methylethyl, ipr2pcl, pubchem6476, chlorodiisopropylphosphane, chloro diisopropylphosphine, acmc-209jc6, diisopropylphosphinyl chloride | |
CC(C)P(C(C)C)Cl | |
152.61 | |
152.61 | |
Liquid |
Chemical Identifiers
40244-90-4 | |
152.61 | |
JZPDBTOWHLZQFC-UHFFFAOYSA-N | |
538967 | |
CC(C)P(C(C)C)Cl |
C6H14ClP | |
MFCD00015027 | |
chlorodiisopropylphosphine, diisopropylchlorophosphine, di-i-propylchlorophosphine, phosphinous chloride, bis 1-methylethyl, ipr2pcl, pubchem6476, chlorodiisopropylphosphane, chloro diisopropylphosphine, acmc-209jc6, diisopropylphosphinyl chloride | |
chloro-di(propan-2-yl)phosphane |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only