missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorodifluoroacetic Acid, TCI America™

$29.03 - $403.34

Chemical Identifiers

CAS 76-04-0
Molecular Formula C2HClF2O2
Molecular Weight (g/mol) 130.475
MDL Number MFCD00004176
InChI Key OAWAZQITIZDJRB-UHFFFAOYSA-N
Synonym chlorodifluoroacetic acid, acetic acid, chlorodifluoro, difluorochloroacetic acid, chloro difluoro acetic acid, 2-chloro-2,2-difluoro-acetic acid, acetic acid, 2-chloro-2,2-difluoro, snpljqdjptadtaetp@, chlorodifluoroaceticacid, chlorodifluoro acetic acid, acmc-209p1k
PubChem CID 66154
IUPAC Name 2-chloro-2,2-difluoroacetic acid
SMILES C(=O)(C(F)(F)Cl)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C099025G View Documents TCI America
C099025G
25g Each for $29.03
 
C0990500G View Documents TCI America
C0990500G
500g Each for $403.34
 
Specifications

Chemical Identifiers

76-04-0
130.475
OAWAZQITIZDJRB-UHFFFAOYSA-N
66154
C(=O)(C(F)(F)Cl)O
C2HClF2O2
MFCD00004176
chlorodifluoroacetic acid, acetic acid, chlorodifluoro, difluorochloroacetic acid, chloro difluoro acetic acid, 2-chloro-2,2-difluoro-acetic acid, acetic acid, 2-chloro-2,2-difluoro, snpljqdjptadtaetp@, chlorodifluoroaceticacid, chlorodifluoro acetic acid, acmc-209p1k
2-chloro-2,2-difluoroacetic acid

Specifications

126°C
25g
C2HClF2O2
3261
OAWAZQITIZDJRB-UHFFFAOYSA-N
2-chloro-2,2-difluoroacetic acid
66154
Crystal-Lump at 20°C
Chlorodifluoroacetic Acid
Undesignated
76-04-0
MFCD00004176
chlorodifluoroacetic acid, acetic acid, chlorodifluoro, difluorochloroacetic acid, chloro difluoro acetic acid, 2-chloro-2,2-difluoro-acetic acid, acetic acid, 2-chloro-2,2-difluoro, snpljqdjptadtaetp@, chlorodifluoroaceticacid, chlorodifluoro acetic acid, acmc-209p1k
C(=O)(C(F)(F)Cl)O
130.475
130.47
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : Yes