Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorodifluoroacetic acid, 98+%

$142.39 - $142.39

Chemical Identifiers

Chemical Name or Material Chlorodifluoroacetic acid
CAS 76-04-0
Molecular Formula ClCF2CO2H
Formula Weight 130.48
MDL Number MFCD00004176
UN Number UN3265
Beilstein 956625
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1436922

 
alfa aesar™
A1436922
100g Each for $142.39

AAA1436914

 
alfa aesar™
A1436914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.48
Chemical Name or Material Chlorodifluoroacetic acid
TSCA TSCA
Odor Acrid
Assay ≥98%
Molecular Formula ClCF2CO2H
MDL Number MFCD00004176
CAS 76-04-0
UN Number UN3265
Beilstein 956625
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 121° to 122°C
Melting Point 20° to 24°C
Quantity 100g

Applications
It is employed as a starting material for generating difluorocarbene. It is also used as a pharmaceutical and agrochemicals intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, bases, active metals and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.