missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorodicyclohexylphosphine, 97%, Thermo Scientific Chemicals

$109.01 - $109.01

Chemical Identifiers

CAS 16523-54-9
Molecular Formula C12H22ClP
Molecular Weight (g/mol) 232.73
MDL Number MFCD00014294
InChI Key AKJFBIZAEPTXIL-UHFFFAOYSA-N
Synonym chlorodicyclohexylphosphine, dicyclohexylchlorophosphine, dicyclohexylphosphinous chloride, dicyclohexylphosphine chloride, chlorodicyclohexyl phosphine, phosphinous chloride, dicyclohexyl, clpcy2, pcy2cl, chlorodicyclohexvlphosphine, dicyclohexylchlorophosphane
PubChem CID 2734135
IUPAC Name chloro(dicyclohexyl)phosphane
SMILES ClP(C1CCCCC1)C1CCCCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC353290010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
353290010
1 g Glass bottle
Each for $109.01
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16523-54-9
232.73
AKJFBIZAEPTXIL-UHFFFAOYSA-N
2734135
ClP(C1CCCCC1)C1CCCCC1
C12H22ClP
MFCD00014294
chlorodicyclohexylphosphine, dicyclohexylchlorophosphine, dicyclohexylphosphinous chloride, dicyclohexylphosphine chloride, chlorodicyclohexyl phosphine, phosphinous chloride, dicyclohexyl, clpcy2, pcy2cl, chlorodicyclohexvlphosphine, dicyclohexylchlorophosphane
chloro(dicyclohexyl)phosphane

Specifications

16523-54-9
1.0540g/mL
>110°C
96% min. (ex Cl) (Argentometry)
C12H22ClP
(C6H11)2PCl
1 g
chlorodicyclohexylphosphine, dicyclohexylchlorophosphine, dicyclohexylphosphinous chloride, dicyclohexylphosphine chloride, chlorodicyclohexyl phosphine, phosphinous chloride, dicyclohexyl, clpcy2, pcy2cl, chlorodicyclohexvlphosphine, dicyclohexylchlorophosphane
AKJFBIZAEPTXIL-UHFFFAOYSA-N
chloro(dicyclohexyl)phosphane
2734135
97%
Chlorodicyclohexylphosphine
Colorless
165.0°C (12.0 mmHg)
Authentic
Glass bottle
1.5310 to 1.5340
MFCD00014294
1.054
Solubility in water: reacts violently with water
ClP(C1CCCCC1)C1CCCCC1
232.73
232.74
Liquid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only