missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorodi(p-tolyl)phosphine, TCI America™

Manufacturer:  TCI America C32011G

 View more versions of this product

Catalog No. C32011G



Specifications

Specifications

Chlorodi(p-tolyl)phosphine
Yellow
1019-71-2
MFCD01630844
bis 4-methylphenyl chlorophosphine, chlorodi p-tolyl phosphine, chlorodi-p-tolylphosphine, chlorobis 4-methylphenyl phosphine, bis 4-methylphenyl phosphinous chloride, chlorobis 4-methylphenyl phosphane, chloro-bis 4-methylphenyl phosphane, di-p-tolylchlorophosphine, chloro bis-p-tolyl phosphane, di p-tolyl phosphinous chloride
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)Cl
248.69
248.69
≥98.0% (T)
350°C
1g
C14H14ClP
3265
BJBXRRHIBSXGLF-UHFFFAOYSA-N
chloro-bis(4-methylphenyl)phosphane
4188608
Clear Liquid at 20°C

Chemical Identifiers

1019-71-2
248.69
BJBXRRHIBSXGLF-UHFFFAOYSA-N
4188608
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)Cl
C14H14ClP
MFCD01630844
bis 4-methylphenyl chlorophosphine, chlorodi p-tolyl phosphine, chlorodi-p-tolylphosphine, chlorobis 4-methylphenyl phosphine, bis 4-methylphenyl phosphinous chloride, chlorobis 4-methylphenyl phosphane, chloro-bis 4-methylphenyl phosphane, di-p-tolylchlorophosphine, chloro bis-p-tolyl phosphane, di p-tolyl phosphinous chloride
chloro-bis(4-methylphenyl)phosphane
Safety and Handling

Safety and Handling

TSCA : No

Documents