Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorodi(o-tolyl)phosphine, 98%

Chemical Identifiers

Chemical Name or Material Chlorodi(o-tolyl)phosphine
CAS 36042-94-1
Molecular Formula C14H14ClP
Formula Weight 248.69
MDL Number MFCD04038733
UN Number UN1760
Synonym Di(o-tolyl)chlorophosphine; Chlorobis(2-methylphenyl)phosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2589003

 
alfa aesar™
H2589003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2589006

 
alfa aesar™
H2589006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 248.69
Chemical Name or Material Chlorodi(o-tolyl)phosphine
Assay 98%
Molecular Formula C14H14ClP
MDL Number MFCD04038733
CAS 36042-94-1
UN Number UN1760
Synonym Di(o-tolyl)chlorophosphine; Chlorobis(2-methylphenyl)phosphine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Boiling Point 174° to 178°C (3mmHg)
Melting Point 57°C
Quantity 1g

Applications
Chlorodi(o-tolyl)phosphine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Not miscible or difficult to mix in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.