Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorodi(1-naphthyl)phosphine, 95%

Chemical Identifiers

Chemical Name or Material Chlorodi(1-naphthyl)phosphine
CAS 36042-99-6
Molecular Formula C20H14ClP
Formula Weight 320.76
MDL Number MFCD09753469
UN Number UN3261
Synonym Di(1-naphthyl)chlorophosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2589303

 
alfa aesar™
H2589303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2589306

 
alfa aesar™
H2589306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 320.76
Chemical Name or Material Chlorodi(1-naphthyl)phosphine
Assay 95%
Molecular Formula C20H14ClP
MDL Number MFCD09753469
CAS 36042-99-6
UN Number UN3261
Synonym Di(1-naphthyl)chlorophosphine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 227° to 229°C (0.6mmHg)
Melting Point 154° to 157°C
Quantity 1g

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.