Please login to your online account to display your discounted pricing

Chlorocyclopentadienylbis(triphenylphosphine)ruthenium(II), 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Chlorocyclopentadienylbis(triphenylphosphine)ruthenium(II)
Assay Percent Range 98%
Molecular Formula C41H35ClP2Ru
Formula Weight 726.2
MDL Number MFCD00075004
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC377880010

  SDS
thermo fisher scientific
AC377880010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 726.2
Quantity 1g
Color Orange
Ruthenium (Ru) 97.5% min. (based on Ru)
Molecular Formula C41H35ClP2Ru
Packaging Glass bottle
Assay Percent Range 98%
MDL Number MFCD00075004
CAS 32993-05-8
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Chlorocyclopentadienylbis(triphenylphosphine)ruthenium(II)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.