Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorocyclobutane, 97%

Chemical Identifiers

Chemical Name or Material Chlorocyclobutane
CAS 1120-57-6
Molecular Formula C4H7Cl
Formula Weight 90.55
MDL Number MFCD00013736
UN Number UN1127
Beilstein 1900414
Synonym Cyclobutyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1256103

  SDS
alfa aesar™
A1256103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1256106

  SDS
alfa aesar™
A1256106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 90.55
UN Number UN1127
Odor Slightly Sweetish
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1900414
Molecular Formula C4H7Cl
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES
Flash Point −8°C (17°F)
Boiling Point 80° to 81°C
CAS 1120-57-6
Health Hazard 1 Danger
MDL Number MFCD00013736
Assay 97%
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Quantity 1g
Chemical Name or Material Chlorocyclobutane
Synonym Cyclobutyl chloride

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Insoluble in water.

Notes
Store at 4°C. Protect against electrostatic charges.

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.