Please login to your online account to display your discounted pricing

α-chlorocinnamaldehyde, +90.5%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC393531000

 View more versions of this product

Catalog No. AC393531000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid After Melting
Assay ≥90.0%
Formula Weight 166.61
Quantity 100g
Color Brown
CAS Min % 90.0
CAS Max % 100.0
Chemical Name or Material α-chlorocinnamaldehyde
Linear Formula C6H5CH=C(Cl)CHO
Infrared Spectrum Authentic
Assay Percent Range 90+%
Molecular Formula C9H7ClO
MDL Number MFCD00077440
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
CAS 18365-42-9
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material α-chlorocinnamaldehyde
CAS 18365-42-9
CAS Min % 90.0
CAS Max % 100.0
Assay ≥90.0%
Assay Percent Range 90+%
Molecular Formula C9H7ClO
Linear Formula C6H5CH=C(Cl)CHO
Formula Weight 166.61
MDL Number MFCD00077440
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents