Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorocarbonylsulfenyl chloride, 97%

$99.30 - $99.30

Chemical Identifiers

Chemical Name or Material Chlorocarbonylsulfenyl chloride
CAS 2757-23-5
Molecular Formula CCl2OS
Formula Weight 130.98
MDL Number MFCD00000703
UN Number UN3265
Beilstein 506318
Synonym Chloroformylsulfenyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0643203

  SDS
alfa aesar™
L0643203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0643206

  SDS
alfa aesar™
L0643206
5g Each for $99.30
Description & Specifications

Specifications

Formula Weight 130.98
Chemical Name or Material Chlorocarbonylsulfenyl chloride
MDL Number MFCD00000703
Assay 97%
CAS 2757-23-5
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym Chloroformylsulfenyl chloride
Molecular Formula CCl2OS
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 1g
Boiling Point 98° to 101°C
Health Hazard 1 Danger
Beilstein 506318
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
UN Number UN3265
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.