missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorocarbonylsulfenyl chloride, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC224340250

 View more versions of this product

Catalog No. AC224340250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Chlorocarbonylsulfenyl chloride
Yellow
2757-23-5
Glass bottle
≥94%
CCl2OS
130.98
1.5800g/mL
25g
Authentic
Liquid, Fuming In Moist Air
95%
ClCOSCl
MFCD00000703

Chemical Identifiers

95%
ClCOSCl
MFCD00000703
≥94%
CCl2OS
130.98
Chlorocarbonylsulfenyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger