Please login to your online account to display your discounted pricing

Chlorocarbonyl-1-methylethyl acetate, 90%


Manufacturer: thermo fisher scientific  AC271430250

 View more versions of this product

Catalog No. AC271430250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 164.59
Physical Form Liquid
Assay ≥88%
Color Clear colorless
Quantity 25g
Density 1.1300g/mL
UN Number 3265
Linear Formula CH3C(O)OC(CH3)2C(O)Cl
Chemical Name or Material Chlorocarbonyl-1-methylethyl acetate
Assay Percent Range 90%
Molecular Formula C6H9ClO3
MDL Number MFCD00000708
CAS 40635-66-3
Synonym 1-Chlorocarbonyl-1-methylethyl acetate
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
NMR Consistent with structure

Specifications

Chemical Name or Material Chlorocarbonyl-1-methylethyl acetate
CAS 40635-66-3
Molecular Formula C6H9ClO3
Linear Formula CH3C(O)OC(CH3)2C(O)Cl
Formula Weight 164.59
MDL Number MFCD00000708
UN Number 3265
Synonym 1-Chlorocarbonyl-1-methylethyl acetate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.