missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorocarbonyl-1-methylethyl acetate, 90%

Manufacturer:  Thermo Fisher Scientific AC271430050

 View more versions of this product

Catalog No. AC271430050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

5g
Liquid
164.59
3265
40635-66-3
C6H9ClO3
Chlorocarbonyl-1-methylethyl acetate
Glass bottle
Consistent with structure
Clear colorless
≥88%
1.1300g/mL
1-Chlorocarbonyl-1-methylethyl acetate
90%
MFCD00000708
CH3C(O)OC(CH3)2C(O)Cl
Authentic

Chemical Identifiers

40635-66-3
CH3C(O)OC(CH3)2C(O)Cl
MFCD00000708
1-Chlorocarbonyl-1-methylethyl acetate
C6H9ClO3
164.59
3265
Chlorocarbonyl-1-methylethyl acetate

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger