Please login to your online account to display your discounted pricing

Chlorocarbonyl-1-methylethyl acetate, 90%

Chemical Identifiers

Chemical Name or Material Chlorocarbonyl-1-methylethyl acetate
CAS 40635-66-3
Molecular Formula C6H9ClO3
Linear Formula CH3C(O)OC(CH3)2C(O)Cl
Formula Weight 164.59
MDL Number MFCD00000708
UN Number 3265
Synonym 1-Chlorocarbonyl-1-methylethyl acetate
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AC271430250

 
thermo fisher scientific
AC271430250
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC271430050

  SDS
thermo fisher scientific
AC271430050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.59
Physical Form Liquid
Assay ≥88%
Color Clear colorless
Quantity 25g
Density 1.1300g/mL
UN Number 3265
Linear Formula CH3C(O)OC(CH3)2C(O)Cl
Synonym 1-Chlorocarbonyl-1-methylethyl acetate
MDL Number MFCD00000708
CAS 40635-66-3
Chemical Name or Material Chlorocarbonyl-1-methylethyl acetate
Assay Percent Range 90%
Molecular Formula C6H9ClO3
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
NMR Consistent with structure
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.