Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorobis(ethylene)rhodium(I) dimer, 99%

Chemical Identifiers

Chemical Name or Material Chlorobis(ethylene)rhodium(I) dimer
CAS 12081-16-2
Formula Weight 388.93
MDL Number MFCD00013206
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3929103

  SDS
alfa aesar™
3929103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 388.93
Chemical Name or Material Chlorobis(ethylene)rhodium(I) dimer
Physical Form Powder
Assay 99%
MDL Number MFCD00013206
CAS 12081-16-2
Sensitivity Air Sensitive
Quantity 1g