missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorobis(cyclooctene)iridium(I) dimer, Ir nominally 42.9%

Manufacturer:  Alfa Aesar™ 4496603

 View more versions of this product

Catalog No. AA4496603



Specifications

Specifications

Chlorobis (cyclooctene)iridium (I) dimer
12246-51-4
MFCD00213465
Ir nominally 42.9%
C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.[Cl-].[Cl-].[Ir].[Ir]
896.134
896
1g
C32H56Cl2Ir2-2
chlorobis cyclooctene iridium i dimer, di-mu-chlorotetrakis cyclooctene diiridium i, bis cyclooctene iridium i chloride, dimer, chlorobis cyclooctene iridium i dimer, chlorobis cyclooctene-iridium i dimer, chlorobis cyclooctene iridium dimer, di-mu-chlorobis cyclooctene iridium i
CJJIQMGSHWWMCK-XFCUKONHSA-L
cyclooctene;iridium;dichloride
91972094

Chemical Identifiers

12246-51-4
896.134
CJJIQMGSHWWMCK-XFCUKONHSA-L
91972094
C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.[Cl-].[Cl-].[Ir].[Ir]
C32H56Cl2Ir2-2
MFCD00213465
chlorobis cyclooctene iridium i dimer, di-mu-chlorotetrakis cyclooctene diiridium i, bis cyclooctene iridium i chloride, dimer, chlorobis cyclooctene iridium i dimer, chlorobis cyclooctene-iridium i dimer, chlorobis cyclooctene iridium dimer, di-mu-chlorobis cyclooctene iridium i
cyclooctene;iridium;dichloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents