Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorobis(4-methoxyphenyl)phosphine, 98+%

$147.35 - $442.99

Chemical Identifiers

Chemical Name or Material Chlorobis(4-methoxyphenyl)phosphine
CAS 13685-30-8
Molecular Formula C14H14ClO2P
Formula Weight 280.69
MDL Number MFCD04972301
UN Number UN3261
Synonym Bis(4-methoxyphenyl)chlorophosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2588703

  SDS
alfa aesar™
H2588703
1g Each for $147.35

AAH2588706

  SDS
alfa aesar™
H2588706
5g Each for $442.99
Description & Specifications

Specifications

Formula Weight 280.69
Chemical Name or Material Chlorobis(4-methoxyphenyl)phosphine
Odor Irritating
Assay ≥98%
Molecular Formula C14H14ClO2P
MDL Number MFCD04972301
CAS 13685-30-8
UN Number UN3261
Synonym Bis(4-methoxyphenyl)chlorophosphine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 140°C (1.5mmHg)
Melting Point 61° to 64°C
Quantity 1g

Applications
Chlorobis(4-methoxyphenyl)phosphine is used an pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Air & moisture sensitive. Store away from oxidizing agents, bases, air, water/ moisture.

Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.