Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorobis(4-fluorophenyl)phosphine, 98%

Chemical Identifiers

Chemical Name or Material Chlorobis(4-fluorophenyl)phosphine
CAS 23039-97-6
Molecular Formula C12H8ClF2P
Formula Weight 256.62
MDL Number MFCD04972302
UN Number UN3265
Synonym Bis(4-fluorophenyl)chlorophosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2695303

  SDS
alfa aesar™
H2695303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2695306

  SDS
alfa aesar™
H2695306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH26953MD

  SDS
alfa aesar™
H26953MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 256.62
Assay 98%
Chemical Name or Material Chlorobis(4-fluorophenyl)phosphine
Molecular Formula C12H8ClF2P
MDL Number MFCD04972302
CAS 23039-97-6
UN Number UN3265
Synonym Bis(4-fluorophenyl)chlorophosphine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Quantity 1g

Applications
In chemical research. Used as an interphasic transfer catalyst, a polymerization catalyst, and a trans-halogenation catalyst. As a precursor to fire-retardant materials.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.