Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorobis(4-chlorophenyl)phosphine, 97%

Chemical Identifiers

Chemical Name or Material Chlorobis(4-chlorophenyl)phosphine
CAS 13685-26-2
Molecular Formula C12H8Cl3P
Formula Weight 289.53
MDL Number MFCD09909564
UN Number UN3261
Synonym Bis(4-chlorophenyl)chlorophosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2588603

 
alfa aesar™
H2588603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2588606

 
alfa aesar™
H2588606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 289.53
Chemical Name or Material Chlorobis(4-chlorophenyl)phosphine
Assay 97%
Molecular Formula C12H8Cl3P
MDL Number MFCD09909564
CAS 13685-26-2
UN Number UN3261
Synonym Bis(4-chlorophenyl)chlorophosphine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Boiling Point 184° to 186°C (2.5mmHg)
Melting Point 52° to 53°C
Quantity 1g

Applications
Chlorobis(4-chlorophenyl)phosphine are now employed in tuning the properties of catalyst. Also used as primary and secondary intermediates.

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.