Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorobis(3,5-dimethylphenyl)phosphine, tech. 90%

$123.41 - $123.41

Chemical Identifiers

Chemical Name or Material Chlorobis(3,5-dimethylphenyl)phosphine
CAS 74289-57-9
Purity Grade Technical Grade
Molecular Formula C16H18ClP
Formula Weight 276.75
MDL Number MFCD01630841
UN Number UN3261
Synonym Bis(3,5-dimethylphenyl)chlorophosphine; Chlorodi(3,5-xylyl)phosphine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2588503

  SDS
alfa aesar™
H2588503
1g Each for $123.41

AAH2588506

  SDS
alfa aesar™
H2588506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 276.75
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Assay 90%
CAS 74289-57-9
Synonym Bis(3,5-dimethylphenyl)chlorophosphine; Chlorodi(3,5-xylyl)phosphine
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD01630841
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
UN Number UN3261
Chemical Name or Material Chlorobis(3,5-dimethylphenyl)phosphine
Purity Grade Technical Grade
Molecular Formula C16H18ClP

Applications
Chlorobis(3,5-dimethylphenyl)phosphine is used in the synthesis of (S)- and (R)-2,2′-Bis[bis(3,5-dimethylphenyl)phosphinoyl]-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-binaphthyl (Xyl-H 8 -BINAPO).

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.