Please login to your online account to display your discounted pricing

Chlorobenzene (Laboratory), Fisher Chemical


Quantity 20L
Packaging Steel Pail

Manufacturer: fisher scientific company  BPB25420

 View more versions of this product

Catalog No. B254-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Melting Point −45°C
Boiling Point 131°C
Formula Weight 112.56
Purity Grade Laboratory Grade
Quantity 20L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Clear
Density 1.108g/cm³
Vapor Pressure 12mbar at 20°C
Viscosity 0.8 mPa/s at 20°C
Vapor Density 3.9g/mol
UN Number 1134
Identification Pass Test
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes and skin. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:3
Flammability:3
Instability:0
MDL Number MFCD00000530
Additional Information Vapor Pressure: 12mbar at 20°C
Packaging Steel Pail
Synonym Benzene Chloride
Health Hazard 2 WARNING!
ChemAlert Storage Symbol Red
Cleanup Symbol Solvent
CAS 108-90-7
Chemical Name or Material Chlorobenzene
Molecular Formula C6H5Cl

Specifications

Chemical Name or Material Chlorobenzene
CAS 108-90-7
Purity Grade Laboratory Grade
Molecular Formula C6H5Cl
Formula Weight 112.56
MDL Number MFCD00000530
Synonym Benzene Chloride
Accessories
 

Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes and skin. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:3
Flammability:3
Instability:0

WARNING!

 

SDS & Safety Documents