missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorobenzene, HPLC Grade, 99.5%

Manufacturer:  Alfa Aesar™ 22922K2

 View more versions of this product

Catalog No. AA22922K2



Specifications

Specifications

Chlorobenzene
HPLC Grade
C6H5Cl
UN1134
14,2121
C1=CC=C(C=C1)Cl
112.556
CHEBI:28097
Liquid
Aromatic
28° (82°F)
Colorless
1L
108-90-7
99.5%
MFCD00000530
605632
MVPPADPHJFYWMZ-UHFFFAOYSA-N
chlorobenzene
7964
112.56
1.106g/mL
1.5240
132°C
-45°C

Chemical Identifiers

108-90-7
112.556
MVPPADPHJFYWMZ-UHFFFAOYSA-N
CHEBI:28097
C1=CC=C(C=C1)Cl
C6H5Cl
MFCD00000530
7964
chlorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE

RTECSNumber : CZ0175000

TSCA : TSCA

SDS Documents