Learn More
Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific Chemicals
$162.84 - $519.85
Chemical Identifiers
CAS | 108-90-7 |
---|---|
Molecular Formula | C6H5Cl |
Molecular Weight (g/mol) | 112.556 |
MDL Number | MFCD00000530 |
InChI Key | MVPPADPHJFYWMZ-UHFFFAOYSA-N |
Synonym | monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene |
PubChem CID | 7964 |
ChEBI | CHEBI:28097 |
IUPAC Name | chlorobenzene |
SMILES | C1=CC=C(C=C1)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA22922K2
|
Thermo Scientific Chemicals
022922K2 |
1 L |
Each for $162.84
|
|
|||||
AA22922M1
|
Thermo Scientific Chemicals
022922M1 |
4 x 1 L |
Each for $519.85
|
|
|||||
AA22922K7
|
Thermo Scientific Chemicals
022922K7 |
4 L |
Each for $504.42
|
|
|||||
Description
Chlorobenzene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-90-7 | |
112.556 | |
MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
7964 | |
chlorobenzene |
C6H5Cl | |
MFCD00000530 | |
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
CHEBI:28097 | |
C1=CC=C(C=C1)Cl |
Specifications
-45°C | |
Colorless | |
132°C | |
28°C (82°F) | |
Liquid | |
C6H5Cl | |
UN1134 | |
14,2121 | |
Miscible with methanol and diethyl ether. Immiscible with water. | |
C1=CC=C(C=C1)Cl | |
112.556 | |
CHEBI:28097 | |
99.5% | |
Chlorobenzene |
108-90-7 | |
1.106 g/mL | |
1 L | |
Aromatic | |
1.524 | |
MFCD00000530 | |
605632 | |
monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene | |
MVPPADPHJFYWMZ-UHFFFAOYSA-N | |
chlorobenzene | |
7964 | |
112.56 | |
HPLC |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
H226-H315-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE
EINECSNumber : 203-628-5
RTECSNumber : CZ0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only