Please login to your online account to display your discounted pricing

Chlorobenzene, EMD Millipore

For synthesis

$198.00 - $198.00

Chemical Identifiers

Chemical Name or Material Chlorobenzene
CAS 108-90-7
Molecular Formula C6H5Cl
Formula Weight 112.56
UN Number 1134
Synonym Phenyl chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M8017912500

 
emd millipore corporation
8017912500
Glass Bottle 2.5L Each for $198.00
Description & Specifications

Specifications

Boiling Point 132°C
Melting Point −45°C
pH Neutral (20°C)
Formula Weight 112.56
Quantity 2.5L
Chemical Name or Material Chlorobenzene
Odor Benzene-like
Packaging Glass Bottle
Refractive Index 1.52
Storage Information 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.
Vapor Pressure 12 hPa (20°C)
Flash Point 27°C (80.6°F)
Physical Form Liquid
Color Colorless
Molecular Formula C6H5Cl
CAS 108-90-7
UN Number 1134
Synonym Phenyl chloride
Solubility Information Soluble in water: 0.4g/L at 20°C
Health Hazard 1 H226: Flammable liquid and vapour.
Health Hazard 2 H315 Causes skin irritation.
Density 1.11g/cm3 at 20°C
Health Hazard 3 H332: Harmful if inhaled.
Safety and Handling

H226: Flammable liquid and vapour.

H332: Harmful if inhaled.

H315 Causes skin irritation.