Please login to your online account to display your discounted pricing

Chlorobenzene-d5, for NMR, 98.5+% atom D, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  175320050

 View more versions of this product

Catalog No. AC175320050

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade For NMR
Physical Form Liquid
Formula Weight 117.6
Quantity 5g
Color Clear colorless
EINECS Number 221-482-0
Molecular Formula C6ClD5
Linear Formula C6D5Cl
CAS 3114-55-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Chlorobenzene-d5
Name Note 98.5+ atom % D
MDL Number MFCD00000531
Deuterium (D) 98.50% min.
Packaging Glass bottle

Specifications

Chemical Name or Material Chlorobenzene-d5
Name Note 98.5+ atom % D
CAS 3114-55-4
Purity Grade For NMR
Molecular Formula C6ClD5
Linear Formula C6D5Cl
Formula Weight 117.6
MDL Number MFCD00000531
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

SDS & Safety Documents