Please login to your online account to display your discounted pricing

Chlorobenzene (Certified), Fisher Chemical


Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPB255500

 View more versions of this product

Catalog No. B255-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Boiling Point 131°C
Melting Point −45°C
Formula Weight 112.56
Quantity 500mL
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 12mbar at 20°C
Viscosity 0.8 mPa/s at 20°C
Physical Form Liquid
Color Clear
Density 1.108g/cm³
Identification Pass Test
Packaging Glass Bottle
Residue after Evaporation 0.02% max.
Water 0.02% max.
MDL Number MFCD00000530
UN Number 1134
Chemical Name or Material Chlorobenzene
Additional Information Vapor Pressure: 12mbar at 20°C
Refractive Index 1.5240 to 1.5250 (at 20°C)
Cleanup Symbol Solvent
ChemAlert Storage Symbol Red
Synonym Benzene Chloride
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes and skin. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:3
Flammability:3
Instability:0
Molecular Formula C6H5Cl
CAS 108-90-7

Specifications

Chemical Name or Material Chlorobenzene
CAS 108-90-7
Purity Grade Certified Grade
Molecular Formula C6H5Cl
Formula Weight 112.56
MDL Number MFCD00000530
Synonym Benzene Chloride
Accessories
 

Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes and skin. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:3
Flammability:3
Instability:0

WARNING!

 

SDS & Safety Documents