Please login to your online account to display your discounted pricing

Chlorobenzene (Certified), Fisher Chemical

$146.31 - $227.15

Chemical Identifiers

Chemical Name or Material Chlorobenzene
CAS 108-90-7
Purity Grade Certified Grade
Molecular Formula C6H5Cl
Formula Weight 112.56
MDL Number MFCD00000530
Synonym Benzene Chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

B255-500

  SDS
fisher scientific company
BPB255500
Glass Bottle 500mL Each for $146.31
Case of 6 Each for $877.86

B255-1

  SDS
fisher scientific company
BPB2551
Glass Bottle 1L Each for $227.15
Case of 6 Each for $1,362.90
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 112.56
Melting Point −45°C
Boiling Point 131°C
Quantity 500mL
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Density 1.108g/cm³
Color Clear
Vapor Pressure 12mbar at 20°C
Viscosity 0.8 mPa/s at 20°C
MDL Number MFCD00000530
UN Number 1134
Residue after Evaporation 0.02% max.
Packaging Glass Bottle
Water 0.02% max.
Identification Pass Test
Refractive Index 1.5240 to 1.5250 (at 20°C)
Additional Information Vapor Pressure: 12mbar at 20°C
Synonym Benzene Chloride
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes and skin. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:3
Flammability:3
Instability:0
ChemAlert Storage Symbol Red
Cleanup Symbol Solvent
Chemical Name or Material Chlorobenzene
CAS 108-90-7
Molecular Formula C6H5Cl
Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. Harmful by inhalation. Irritating to eyes and skin. May cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:3
Flammability:3
Instability:0

WARNING!